MilliporeSigma
  • Concise synthesis and biological evaluation of 2-Aroyl-5-amino benzo[b]thiophene derivatives as a novel class of potent antimitotic agents.
Concise synthesis and biological evaluation of 2-Aroyl-5-amino benzo[b]thiophene derivatives as a novel class of potent antimitotic agents.
Journal of medicinal chemistry (2013-10-30)
Romeo Romagnoli, Pier Giovanni Baraldi, Carlota Lopez-Cara, Delia Preti, Mojgan Aghazadeh Tabrizi, Jan Balzarini, Marcella Bassetto, Andrea Brancale, Xian-Hua Fu, Yang Gao, Jun Li, Su-Zhan Zhang, Ernest Hamel, Roberta Bortolozzi, Giuseppe Basso, Giampietro Viola
ABSTRACT
The biological importance of microtubules make them an interesting target for the synthesis of antitumor agents. The 2-(3',4',5'-trimethoxybenzoyl)-5-aminobenzo[b]thiophene moiety was identified as a novel scaffold for the preparation of potent inhibitors of microtubule polymerization acting through the colchicine site of tubulin. The position of the methoxy group on the benzo[b]thiophene was important for maximal antiproliferative activity. Structure-activity relationship analysis established that the best activities were obtained with amino and methoxy groups placed at the C-5 and C-7 positions, respectively. Compounds 3c-e showed more potent inhibition of tubulin polymerization than combretastatin A-4 and strong binding to the colchicine site. These compounds also demonstrated substantial antiproliferative activity, with IC50 values ranging from 2.6 to 18 nM in a variety of cancer cell lines. Importantly, compound 3c (50 mg/kg), significantly inhibited the growth of the human osteosarcoma MNNG/HOS xenograft in nude mice.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, suitable for plant cell culture, BioReagent, ≥95% (HPLC)