MilliporeSigma
  • Randomized controlled trial of alfacalcidol supplementation for the reduction of hypocalcemia after total thyroidectomy.
Randomized controlled trial of alfacalcidol supplementation for the reduction of hypocalcemia after total thyroidectomy.
American journal of surgery (2013-10-15)
Laurent Genser, Christophe Trésallet, Gaëlle Godiris-Petit, Stéphanie Li Sun Fui, Harika Salepcioglu, Catherine Royer, Fabrice Menegaux
ABSTRACT
The aim of this study was to evaluate the effect of perioperative alfacalcidol on postoperative hypocalcemia after total thyroidectomy. A total of 219 patients scheduled for total thyroidectomy were randomized into groups not receiving (group A) or receiving (group B) perioperative alfacalcidol. Postoperative hypocalcemia was compared between groups on postoperative day (POD) 1 and POD2. Patients with hypocalcemia (<2.00 mmol/L) received oral calcium supplementation. Calcium and vitamin D levels were measured at 5-week and 6-month follow-ups. The incidence of symptomatic hypocalcemia was significantly lower in group A (P = .02), whereas similarly low levels of calcemia were observed in both groups on POD1 (37% and 30%, respectively; P = not significant) and persisted on POD2 (14% and 6%, respectively; P = not significant). Patients with severe hypocalcemia (<1.90 mmol/L) showed faster recovery in group A compared with group B (6% vs 1%, P = .04). At 5 weeks, calcium and vitamin D levels were similar between the groups. Six months after surgery, 4% (group A) versus 0% (group B) of subjects exhibited permanent hypoparathyroidism (P = .04). Although the treatment did not correct vitamin D deficiency, perioperative alfacalcidol uptake resulted in decreased transient hypocalcemia and related symptoms in patients undergoing total thyroidectomy.
Sigma-Aldrich
1α-Hydroxyvitamin D3, ≥97.0% (HPLC)
Alfacalcidol, European Pharmacopoeia (EP) Reference Standard