MilliporeSigma
  • Application of slightly acidic electrolyzed water for inactivating microbes in a layer breeding house.
Application of slightly acidic electrolyzed water for inactivating microbes in a layer breeding house.
Poultry science (2013-09-21)
X X Hao, B M Li, C Y Wang, Q Zhang, W Cao
ABSTRACT
Lots of microorganisms exist in layer houses can cause bird diseases and worker health concerns. Spraying chemical disinfectants is an effective way to decontaminate pathogenic microorganisms in the air and on surfaces in poultry houses. Slightly acidic electrolyzed water (SAEW, pH 5.0-6.5) is an ideal, environmentally friendly broad-spectrum disinfectant to prevent and control bacterial or viral infection in layer farms. The purpose of this work was to investigate the cleaning effectiveness of SAEW for inactivating the microbes in layer houses. The effect of SAEW was evaluated by solid materials and surface disinfection in a hen house. Results indicate that SAEW with an available chlorine concentration of 250 mg/L, pH value of 6.19, and oxygen reduction potential of 974 mV inactivated 100% of bacteria and fungi in solid materials (dusts, feces, feather, and feed), which is more efficient than common chemical disinfectant such as benzalkonium chloride solution (1:1,000 vol/vol) and povidone-iodine solution (1:1,000 vol/vol). Also, it significantly reduced the microbes on the equipment or facility surfaces (P < 0.05), including floor, wall, feed trough, and water pipe surfaces. Moreover, SAEW effectively decreased the survival rates of Salmonella and Escherichia coli by 21 and 16 percentage points. In addition, spraying the target with tap water before disinfection plays an important role in spray disinfection.
Sigma-Aldrich
Poly(vinylpyrrolidone)–Iodine complex