MilliporeSigma
  • Ugi multicomponent reaction product: the inhibitive effect on DNA oxidation depends upon the isocyanide moiety.
Ugi multicomponent reaction product: the inhibitive effect on DNA oxidation depends upon the isocyanide moiety.
The Journal of organic chemistry (2013-08-27)
Rui Wang, Zai-Qun Liu
ABSTRACT
The hydroxyl-substituted benzoic acid (as phenyl group A in the product), aniline (as phenyl group B in the product), benzaldehyde (as phenyl group C in the product), and four isocyanides are employed to synthesize bis-amide via an Ugi four-component reaction. The effects of the obtained 20 bis-amides on quenching radicals and inhibiting DNA oxidation are estimated. It is found that the antioxidant effectiveness of bis-amide generated by hydroxyl groups is markedly influenced by the structural feature derived from isocyanide. The phenolic hydroxyl group attaching to phenyl group A plays a major role in scavenging radicals, and the radical-scavenging property is reinforced by the structural moiety introduced from ferrocenylmethyl isocyanide. The same conclusion is also obtained when bis-amides are used to inhibit DNA oxidation. It is still found that the ferrocenylmethyl moiety enhances the antioxidant effect of hydroxyl group at phenyl group A in protecting DNA against the oxidation. Moreover, when the bis-amide is prepared by the same isocyanide, e.g. ethyl isocyanoacetate, it is found that the hydroxyl group at phenyl group C plays the major role in inhibiting DNA oxidation, followed by the hydroxyl groups attaching to phenyl groups B and A.
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Supelco
Aniline, analytical standard
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol