MilliporeSigma
  • Regulation of intraocular pressure in mice: structural analysis of dopaminergic and serotonergic systems in response to cabergoline.
Regulation of intraocular pressure in mice: structural analysis of dopaminergic and serotonergic systems in response to cabergoline.
Biochemical pharmacology (2013-08-22)
Chiara Bianca Maria Platania, Gian Marco Leggio, Filippo Drago, Salvatore Salomone, Claudio Bucolo
ABSTRACT
Elevated intraocular pressure (IOP) is the main recognized risk factor of glaucoma. To investigate the contribution of dopaminergic and serotonergic systems in IOP regulation, we used cabergoline, a mixed dopamine and serotonin agonist, in C57BL/6J WT and dopamine D₃ receptor knock-out (D₃R⁻/⁻) mice with normal eye pressure or steroid-induced ocular hypertension. Furthermore, we studied the structural basis of the cabergoline-mediated activation of the dopaminergic and serotonergic systems by molecular modeling. Topical application of cabergoline, significantly decreased, in a dose-dependent manner, the intraocular pressure in WT mice, both in an ocular normotensive group (-9, -5 and -2 mmHg with 5%, 1%, and 0.1%, respectively) and an ocular hypertensive group, with a prolonged effect in this latter group. No change of intraocular pressure was observed after topical application of cabergoline in D₃R⁻/⁻ mice. We modeled and optimized, with molecular dynamics, structures of hD₃, h5HT(1A) and h5HT(2A-C) receptors; thereafter we carried out molecular docking of cabergoline. Docking revealed that binding of cabergoline into D₃ and 5HT(1A) receptors is associated with a better desolvation energy in comparison to 5HT(2A-C) binding. In conclusion, the present study support the hypothesis that dopaminergic system is pivotal to regulate IOP and that D₃R represents an intriguing target in the treatment of glaucoma. Furthermore, the structure-based computational approach adopted in this study is able to build and refine structure models of homologous dopaminergic and serotonergic receptors that may be of interest for structure-based drug discovery of ligands, with dopaminergic selectivity or with multi-pharmacological profile, potentially useful to treat optic neuropathies.
Sigma-Aldrich
Cabergoline, ≥98% (HPLC)
Pricing and availability is not currently available.
Cabergoline, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.