MilliporeSigma
  • Genotoxicity assessment of mouse oocytes by comet assay before vitrification and after warming with three vitrification protocols.
Genotoxicity assessment of mouse oocytes by comet assay before vitrification and after warming with three vitrification protocols.
Fertility and sterility (2013-06-13)
Anais Berthelot-Ricou, Jeanne Perrin, Carole di Giorgio, Michel de Meo, Alain Botta, Blandine Courbiere
ABSTRACT
To assess the genotoxicity of three oocyte vitrification protocols. Murine assay. Biogenotoxicology research laboratory. CD1 female mice. Three mouse oocyte groups were exposed to three commercialized human oocyte vitrification protocols. Protocols 1 and 2 contained dimethyl sulfoxide and ethylene glycol (EG), and protocol 3 contained EG and 1,2-propanediol (PrOH). DNA damage was first evaluated by comet assay after oocyte exposure to the three different equilibration and vitrification solutions. Comet assay was also performed after full vitrification and warming procedure and compared with a negative control group (oocytes stored in medium culture only) and a positive control group (oocytes exposed to hydrogen peroxide just before comet assay). DNA damage was quantified as Olive tail moment (OTM). Statistical analysis consisted of a Shapiro-Wilk test. Then, median protocol OTM was compared with the negative control group with the Mann-Whitney U test. The difference was considered to be statistically significant if the P value was <.05. In both parts of our study, protocols 1 and 2 did not induce significant DNA damage, whereas protocol 3 induced statistically higher DNA damage compared with the negative control group. Vitrification protocols containing PrOH induced significant DNA damage on mouse oocytes, both before cooling and after warming. Therefore, for the moment, we prefer vitrification techniques without PrOH while we await more studies on PrOH toxicity and long-term evaluation.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethylene glycol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylene glycol 5 M solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl sulfoxide, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%