MilliporeSigma
  • Phosphorus-nitrogen compounds. Part 23: syntheses, structural investigations, biological activities, and DNA interactions of new N/O spirocyclotriphosphazenes.
Phosphorus-nitrogen compounds. Part 23: syntheses, structural investigations, biological activities, and DNA interactions of new N/O spirocyclotriphosphazenes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-11-23)
Nuran Asmafiliz, Zeynel Kılıç, Zeliha Hayvalı, Leyla Açık, Tuncer Hökelek, Hakan Dal, Yağmur Oner
ABSTRACT
The Schiff base compounds (1 and 2) are synthesized by the condensation reactions of 2-furan-2-yl-methylamine with 2-hydroxy-3-methoxy- and 2-hydroxy-5-methoxy-benzaldehydes and reduced with NaBH(4) to give the new N/O-donor-type ligands (3 and 4). The monospirocyclotriphosphazenes containing 1,3,2-oxazaphosphorine rings (5 and 6) are prepared from the reactions of N(3)P(3)Cl(6) with 3 and 4, respectively. The reactions of 5 and 6 with excess pyrrolidine, morpholine, and 1,4-dioxa-8-azaspiro [4,5] decane (DASD) produce tetrapyrrolidino (5a and 6a), morpholino (5b and 6b), and 1,4-dioxa-8-azaspiro [4,5] deca (5c and 6c) spirocyclotriphosphazenes. The structural investigations of the compounds are examined by (1)H, (13)C, (31)P NMR, DEPT, HSQC, and HMBC techniques. The solid-state structures of 5, 5a, and 6 are determined using X-ray crystallography. The compounds 5a, 5b, 5c, 6a, 6b, and 6c are subjected to antimicrobial activity against six patojen bacteria and two yeast strains. In addition, interactions between these compounds and pBR322 plasmid DNA are presented by agarose gel electrophoresis.