MilliporeSigma
Evolution and protein packaging of small-molecule RNA aptamers.
ACS nano (2011-09-09)
Jolene L Lau, Michael M Baksh, Jason D Fiedler, Steven D Brown, Amanda Kussrow, Darryl J Bornhop, Phillip Ordoukhanian, M G Finn
ABSTRACT
A high-affinity RNA aptamer (K(d) = 50 nM) was efficiently identified by SELEX against a heteroaryldihydropyrimidine structure, chosen as a representative drug-like molecule with no cross reactivity with mammalian or bacterial cells. This aptamer, its weaker-binding variants, and a known aptamer against theophylline were each embedded in a longer RNA sequence that was encapsidated inside a virus-like particle by a convenient expression technique. These nucleoprotein particles were shown by backscattering interferometry to bind to the small-molecule ligands with affinities similar to those of the free (nonencapsidated) aptamers. The system therefore comprises a general approach to the production and sequestration of functional RNA molecules, characterized by a convenient label-free analytical technique.
Sigma-Aldrich
8-Amino-1-octanethiol hydrochloride, 95%