MilliporeSigma
  • Purification and characterization of a lectin from Phaseolus vulgaris cv. (Anasazi beans).
Purification and characterization of a lectin from Phaseolus vulgaris cv. (Anasazi beans).
Journal of biomedicine & biotechnology (2009-04-04)
Arishya Sharma, Tzi Bun Ng, Jack Ho Wong, Peng Lin
ABSTRACT
A lectin has been isolated from seeds of the Phaseolus vulgaris cv. "Anasazi beans" using a procedure that involved affinity chromatography on Affi-gel blue gel, fast protein liquid chromatography (FPLC)-ion exchange chromatography on Mono S, and FPLC-gel filtration on Superdex 200. The lectin was comprised of two 30-kDa subunits with substantial N-terminal sequence similarity to other Phaseolus lectins. The hemagglutinating activity of the lectin was stable within the pH range of 1-14 and the temperature range of 0-80 degrees C. The lectin potently suppressed proliferation of MCF-7 (breast cancer) cells with an IC(50) of 1.3 microM, and inhibited the activity of HIV-1 reverse transcriptase with an IC(50) of 7.6 microM. The lectin evoked a mitogenic response from murine splenocytes as evidenced by an increase in [3H-methyl]-thymidine incorporation. The lectin had no antifungal activity. It did not stimulate nitric oxide production by murine peritoneal macrophages. Chemical modification results indicated that tryptophan was crucial for the hemagglutinating activity of the lectin.
Sigma-Aldrich
Lectin from Phaseolus vulgaris, lyophilized, powder
Pricing and availability is not currently available.