MilliporeSigma
  • Simple and practical derivatization procedure for enhanced detection of carboxylic acids in liquid chromatography-electrospray ionization-tandem mass spectrometry.
Simple and practical derivatization procedure for enhanced detection of carboxylic acids in liquid chromatography-electrospray ionization-tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2010-04-09)
Tatsuya Higashi, Takuya Ichikawa, Shinsuke Inagaki, Jun Zhe Min, Takeshi Fukushima, Toshimasa Toyo'oka
ABSTRACT
A simple and practical derivatization procedure for increasing the detection responses of carboxylic acids in liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) has been developed. 2-Hydrazinopyridine (HP) and 2-picolylamine (PA) rapidly reacted with biologically and clinically important carboxylic acids [chenodeoxycholic acid, glycochenodeoxycholic acid, prostaglandin E2, 2-(-carboxyethyl)-6-hydroxy-2,7,8-trimethylchroman (gamma-CEHC),alpha-lipoic acid, homovanillic acid (HVA) and 5-hydroxyindole-3-acetic acid] in the presence of 2,2'-dipyridyl disulfide and triphenylphosphine. The resulting HP- and PA-derivatives were highly responsive in ESI-MS operating in the positive-ion mode and gave characteristic product ions during MS/MS, which enabled the sensitive detection using selected reaction monitoring. Among the two reagents, PA was of more practical use; the detection responses of the PA-derivatives were increased by 9-158-fold over the intact carboxylic acids and the limits of detection were in the low femtomole range (1.5-5.6 fmol on column). The PA-derivatization was successfully applied to a biological sample analysis; the derivatization followed by LC-ESI-MS/MS enabled the detection of trace amounts of bile acids, gamma-CEHC and HVA in human saliva with a simple pretreatment, small sample volume and short analysis time.
Product Description
Supelco
4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole, derivatization grade (HPLC), LiChropur, ≥95.0% (HPLC)