MilliporeSigma
  • Orally bioavailable nonpeptide vitronectin receptor antagonists containing 2-aminopyridine arginine mimetics.
Orally bioavailable nonpeptide vitronectin receptor antagonists containing 2-aminopyridine arginine mimetics.
Bioorganic & medicinal chemistry letters (1999-07-16)
R M Keenan, W H Miller, L S Barton, W E Bondinell, R D Cousins, D F Eppley, S M Hwang, C Kwon, M A Lago, T T Nguyen, B R Smith, I N Uzinskas, C C Yuan
ABSTRACT
A peptide RGD analog containing a novel 2-aminopyridine arginine mimetic was discovered to have good affinity and selectivity for the vitronectin receptor. Incorporation of the 2-aminopyridine arginine mimetic into the 3-oxo-1,4-benzodiazepine-2-acetic acid integrin antagonist series led to novel and potent nonpeptide vitronectin receptor antagonists with promising levels of oral bioavailability.