MilliporeSigma
  • Synthesis of potential Rho-kinase inhibitors based on the chemistry of an original heterocycle: 4,4-dimethyl-3,4-dihydro-1H-quinolin-2-one.
Synthesis of potential Rho-kinase inhibitors based on the chemistry of an original heterocycle: 4,4-dimethyl-3,4-dihydro-1H-quinolin-2-one.
European journal of medicinal chemistry (2008-01-01)
Marie-Anne Letellier, Jérôme Guillard, Daniel-Henri Caignard, Gilles Ferry, Jean A Boutin, Marie-Claude Viaud-Massuard
ABSTRACT
A new series of substituted 4,4-dimethyl-3,4-dihydro-1H-quinolin-2-one have been prepared via condensation of 3,3-dimethylacryloyl chloride with aniline. Details of synthetic procedures are shown. Our aim was to investigate the potency of our original heterocycle in the inhibition of the Rho-kinase enzyme, known to be of major importance in the cascade reactions leading to arterial hypertension. Biological activity for the seven compounds has been investigated and is presented.
Sigma-Aldrich
3,3-Dimethylacryloyl chloride, 97%, contains 400 ppm phenothiazine as inhibitor
Pricing and availability is not currently available.