MilliporeSigma
  • Kinetics of inactivation of bovine pancreatic ribonuclease A by bromopyruvic acid.
Kinetics of inactivation of bovine pancreatic ribonuclease A by bromopyruvic acid.
The Biochemical journal (1996-11-15)
M H Wang, Z X Wang, K Y Zhao
ABSTRACT
The kinetic theory of substrate reaction during the modification of enzyme activity [Duggleby (1986) J. Theor. Biol. 123, 67-80; Wang and Tsou (1990) J. Theor. Biol. 142, 531-549] has been applied to a study of the inactivation kinetics of ribonuclease A by bromopyruvic acid. The results show that irreversible inhibition belongs to a non-competitive complexing type inhibition. On the basis of the kinetic equation of substrate reaction in the presence of the inhibitor, all microscopic kinetic constants for the free enzyme, the enzyme-substrate complex and the enzyme-product complex have been determined. The non-competitive inhibition type indicates that neither the substrate nor the product affects the binding of bromopyruvic acid to the enzyme and that the ionization state of His-119 may be the same in both the enzyme-substrate and the enzyme-product complexes.
Sigma-Aldrich
Bromopyruvic acid, ≥98.0%