MilliporeSigma
Cadherin 6 is a new RUNX2 target in TGF-β signalling pathway.
PloS one (2013-09-27)
Valentina Sancisi, Greta Gandolfi, Moira Ragazzi, Davide Nicoli, Ione Tamagnini, Simonetta Piana, Alessia Ciarrocchi
ABSTRACT
Modifications in adhesion molecules profile may change the way tumor cells interact with the surrounding microenvironment. The Cadherin family is a large group of transmembrane proteins that dictate the specificity of the cellular interactions. The Cadherin switch that takes place during epithelial-mesenchymal transition (EMT) contributes to loosening the rigid organization of epithelial tissues and to enhancing motility and invasiveness of tumor cells. Recently, we found Cadherin-6 (CDH6, also known as K-CAD) highly expressed in thyroid tumor cells that display mesenchymal features and aggressive phenotype, following the overexpression of the transcriptional regulator Id1. In this work, we explored the possibility that CDH6 is part of the EMT program in thyroid tumors. We demonstrate that CDH6 is a new transforming growth factor-β (TGF-β) target and that its expression is modulated similarly to other EMT mesenchymal markers, both in vitro and in thyroid tumor patients. We show for the first time that CDH6 is expressed in human thyroid carcinomas and that its expression is enhanced at the invasive front of the tumor. Finally, we show that CDH6 is under the control of the transcription factor RUNX2, which we previously described as a crucial mediator of the Id1 pro-invasive function in thyroid tumor cells. Overall, these observations provide novel information on the mechanism of the EMT program in tumor progression and indicate CDH6 as a potential regulator of invasiveness in thyroid tumors.