MilliporeSigma
  • Design, synthesis and brain uptake of LAT1-targeted amino acid prodrugs of dopamine.
Design, synthesis and brain uptake of LAT1-targeted amino acid prodrugs of dopamine.
Pharmaceutical research (2013-10-19)
Lauri Peura, Kalle Malmioja, Kristiina Huttunen, Jukka Leppänen, Miia Hämäläinen, Markus M Forsberg, Mikko Gynther, Jarkko Rautio, Krista Laine
ABSTRACT
Drug delivery to the brain is impeded by the blood-brain barrier (BBB). Here, we attempted to enhance the brain uptake of cationic dopamine by utilizing the large amino acid transporter 1 (LAT1) at the BBB by prodrug approach. Three amino acid prodrugs of dopamine were synthesized and their prodrug properties were examined in vitro. Their LAT1-binding and BBB-permeation were studied using the in situ rat brain perfusion technique. The brain uptake after intravenous administration and the dopamine-releasing ability in the rat striatum after intraperitoneal administration were also determined for the most promising prodrug. All prodrugs underwent adequate cleavage in rat tissue homogenates. The prodrug with phenylalanine derivative as the promoiety had both higher affinity for LAT1 and better brain uptake properties than those with an alkyl amino acid - mimicking promoiety. The phenylalanine prodrug was taken up into the brain after intravenous injection but after intraperitoneal injection the prodrug did not elevate striatal dopamine concentrations above those achieved by corresponding L-dopa treatment. These results indicate that attachment of phenylalanine to a cationic drug via an amide bond from the meta-position of its aromatic ring could be highly applicable in prodrug design for LAT1-mediated CNS-delivery of not only anionic but also cationic polar drugs.