MilliporeSigma
  • Synthesis of [¹¹C]PBR06 and [¹⁸F]PBR06 as agents for positron emission tomographic (PET) imaging of the translocator protein (TSPO).
Synthesis of [¹¹C]PBR06 and [¹⁸F]PBR06 as agents for positron emission tomographic (PET) imaging of the translocator protein (TSPO).
Steroids (2011-07-16)
Min Wang, Mingzhang Gao, Kathy D Miller, Qi-Huang Zheng
ABSTRACT
The translocator protein 18 kDa (TSPO) is an attractive target for molecular imaging of neuroinflammation and tumor progression. [(18)F]PBR06, a fluorine-18 labeled form of PBR06, is a promising PET TSPO radioligand originally developed at NIMH. [(11)C]PBR06, a carbon-11 labeled form of PBR06, was designed and synthesized for the first time. The standard PBR06 was synthesized from 2,5-dimethoxybenzaldehyde in three steps with 71% overall chemical yield. The radiolabeling precursor desmethyl-PBR06 was synthesized from 2-hydroxy-5-methoxybenzaldehyde in five steps with 12% overall chemical yield. The target tracer [(11)C]PBR06 was prepared by O-[(11)C]methylation of desmethyl-PBR06 with [(11)C]CH(3)OTf in CH(3)CN at 80°C under basic condition and isolated by HPLC combined with SPE purification with 40-60% decay corrected radiochemical yield and 222-740 GBq/μmol specific activity at EOB. On the similar grounds, [(18)F]PBR06 was also designed and synthesized. The previously described Br-PBR06 precursor was synthesized from 2,5-dimethoxybenzaldehyde in two steps with 78% overall chemical yield. A new radiolabeling precursor tosyloxy-PBR06, previously undescribed tosylate congener of PBR06, was designed and synthesized from ethyl 2-hydroxyacetate, 4-methylbenzene-1-sulfonyl chloride, and N-(2,5-dimethoxybenzyl)-2-phenoxyaniline in four steps with 50% overall chemical yield. [(18)F]PBR06 was prepared by the nucleophilic substitution of either new tosyloxy-PBR06 precursor or known Br-PBR06 precursor in DMSO at 140°C with K[(18)F]F/Kryptofix 2.2.2 for 15 min and HPLC combined with SPE purification in 20-60% decay corrected radiochemical yield, >99% radiochemical purity, 87-95% chemical purity, and 37-222 GBq/μmol specific activity at EOB. Radiosynthesis of [(18)F]PBR06 using new tosylated precursor gave similar radiochemical purity, and higher specific activity, radiochemical yield and chemical purity in comparison with radiosynthesis using bromine precursor.
Sigma-Aldrich
2-Hydroxy-5-methoxybenzaldehyde, 98%