MilliporeSigma
4-[(2H-Tetra-zol-2-yl)meth-yl]benzonitrile.
Acta crystallographica. Section E, Structure reports online (2008-01-01)
Zheng Xing, Zhi-Rong Qu
ABSTRACT
The title compound, C(9)H(7)N(5), was synthesized by reaction of 4-(bromomethyl)benzonitrile and 2H-tetrazole in the presence of KOH. The relative orientation of the planar tetra-zole ring and the methyl-benzonitrile moiety is (-)-anti-clinal. The crystal packing is dominated by van der Waals inter-actions.
Sigma-Aldrich
2-(Bromomethyl)benzonitrile, ≥97%
Sigma-Aldrich
4-(Bromomethyl)benzonitrile, 99%