MilliporeSigma
  • Preparation and diastereomeric separation of an (S)- and (R)-1-(methoxycarbonyl)tridec-10-yl glucoside derivative, a precursor for a monosaccharide constituent of resin glycosides.
Preparation and diastereomeric separation of an (S)- and (R)-1-(methoxycarbonyl)tridec-10-yl glucoside derivative, a precursor for a monosaccharide constituent of resin glycosides.
Carbohydrate research (2002-06-01)
Shigeru Kobayashi, Jun ichi Furukawa, Tomoko Sakai, Nobuo Sakairi
ABSTRACT
The 6-O-mesyl derivative of phenyl 1-thio-beta-D-glucopyranoside was prepared from D-glucose as a synthetic equivalent of a 6-deoxy-hexosyl donor. Racemic methyl 11-hydroxytetradecanoate (methyl convolvulinolate) was synthesized by Grignard reaction of propylmagnesium bromide with 10-undecenal followed by hydroboration. Both intermediates were coupled by NIS-TfOH-promoted glycosidation to give a mixture of two diasteromeric glucopyranosides, which were separated on a preparative scale by medium pressure chromatography. One of the products was identified as having the natural (S)-configuration by comparison of its 1H NMR spectrum with an authentic sample prepared from the corresponding chiral hydroxyfatty acid.