MilliporeSigma
  • 6-Substituted 6H-dibenzo[c,h][2,6]naphthyridin-5-ones: reversed lactam analogues of ARC-111 with potent topoisomerase I-targeting activity and cytotoxicity.
6-Substituted 6H-dibenzo[c,h][2,6]naphthyridin-5-ones: reversed lactam analogues of ARC-111 with potent topoisomerase I-targeting activity and cytotoxicity.
Bioorganic & medicinal chemistry (2006-01-18)
Shejin Zhu, Alexander L Ruchelman, Nai Zhou, Angela Liu, Leroy F Liu, Edmond J LaVoie
ABSTRACT
6-Substituted 8,9-dimethoxy-2,3-methylenedioxy-6H-dibenzo[c,h][2,6]naphthyridin-5-ones were synthesized and evaluated for topoisomerase I-targeting activity and cytotoxicity. Several of these reversed lactam analogues of ARC-111 exhibited exceptional cytotoxicity with IC50 values ranging from 0.5 to 3.0 nM. In contrast to topotecan, no resistance was observed with several of these reversed lactam analogues in tumor cell lines that overexpressed the efflux transporters MDR1 or BCRP.
Sigma-Aldrich
2-(2-Chloroethyl)-1-methylpyrrolidine hydrochloride, 99%