MilliporeSigma
  • Chiral amplification in macromolecular helicity assisted by noncovalent interaction with achiral amines and memory of the helical chirality.
Chiral amplification in macromolecular helicity assisted by noncovalent interaction with achiral amines and memory of the helical chirality.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-09-17)
Kazuhide Morino, Nobuyuki Watase, Katsuhiro Maeda, Eiji Yashima
ABSTRACT
Poly[(4-carboxyphenyl)acetylene] (poly-1) exhibits an intense induced circular dichroism (ICD) in the UV-visible region upon complexation with excess (R)-1-(1-naphthyl)ethylamine ((R)-2), owing to the formation of a predominantly single-handed helical conformation of the polymer backbone. In the presence of a small amount of (R)-2, poly-1 showed a very weak ICD due to the lack of a single-handed helical conformation. However, we have found that the co-addition of the excess bulky, achiral 1-naphthylmethylamine (5) with a small amount of (R)-2 caused a dramatic increase in the ICD magnitude, comparable to the full ICD induced by excess (R)-2. This indicates that an almost single-handed helix can be induced on poly-1 upon complexation with a small amount of (R)-2 assisted by achiral 5. Furthermore, the induced single-handed helical poly-1 could be successfully memorized by the replacement of (R)-2 and 5 with achiral 2-aminoethanol or n-butylamine.
Sigma-Aldrich
1-Naphthylmethylamine, 97%