MilliporeSigma
  • DXP reductoisomerase: reaction of the substrate in pieces reveals a catalytic role for the nonreacting phosphodianion group.
DXP reductoisomerase: reaction of the substrate in pieces reveals a catalytic role for the nonreacting phosphodianion group.
Biochemistry (2013-03-12)
Svetlana A Kholodar, Andrew S Murkin
ABSTRACT
The role of the nonreacting phosphodianion group of 1-deoxy-d-xylulose-5-phosphate (DXP) in catalysis by DXP reductoisomerase (DXR) was investigated for the reaction of the "substrate in pieces". The truncated substrate 1-deoxy-l-erythrulose is converted by DXR to 2-C-methylglycerol with a kcat/Km that is 10(6)-fold lower than that for DXP. Phosphite dianion was found to be a nonessential activator, providing 3.2 kcal/mol of transition state stabilization for the truncated substrate. These results implicate a phosphate-driven conformational change involving loop closure over the DXR active site to generate an environment poised for catalysis.
Sigma-Aldrich
1-Deoxy-D-xylulose-5-phosphate sodium salt, ≥99.0% (TLC)
Pricing and availability is not currently available.