MilliporeSigma
  • Dyotropic rearrangements of fused tricyclic β-lactones: application to the synthesis of (-)-curcumanolide A and (-)-curcumalactone.
Dyotropic rearrangements of fused tricyclic β-lactones: application to the synthesis of (-)-curcumanolide A and (-)-curcumalactone.
Journal of the American Chemical Society (2012-08-03)
Carolyn A Leverett, Vikram C Purohit, Alex G Johnson, Rebecca L Davis, Dean J Tantillo, Daniel Romo
ABSTRACT
Dyotropic rearrangements of fused, tricyclic β-lactones are described that proceed via unprecedented stereospecific, 1,2-acyl migrations delivering bridged, spiro-γ-butyrolactones. A unique example of this dyotropic process involves a fused bis-lactone possessing both β- and δ-lactone moieties which enabled rapid access to the core structures of curcumanolide A and curcumalactone. Our current mechanistic understanding of the latter dyotropic process, based on computational studies, is also described. Other key transformations in the described divergent syntheses of (-)-curcumanolide A and (-)-curcumalactone from a common intermediate (11 and 12 steps from 2-methyl-1,3-cyclopentanedione, respectively), include a catalytic, asymmetric nucleophile (Lewis base)-catalyzed aldol-lactonization (NCAL) leading to a tricyclic β-lactone, a Baeyer-Villiger oxidation in the presence of a β-lactone, and highly facial-selective and stereocomplementary reductions of an intermediate spirocyclic enoate. The described dyotropic rearrangements significantly alter the topology of the starting tricyclic β-lactone, providing access to complex spirocyclic cyclopentyl-γ-lactones and bis-γ-lactones in a single synthetic operation.
Sigma-Aldrich
2-Methyl-1,3-cyclopentanedione, 99%
Sigma-Aldrich
(2R)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole, ≥95%