MilliporeSigma
  • Triamino derivatives of triazolotriazine and triazolopyrimidine as adenosine A2a receptor antagonists.
Triamino derivatives of triazolotriazine and triazolopyrimidine as adenosine A2a receptor antagonists.
Bioorganic & medicinal chemistry letters (2004-09-03)
Chi B Vu, Pamela Shields, Bo Peng, Gnanasambandam Kumaravel, Xiaowei Jin, Deepali Phadke, Joy Wang, Thomas Engber, Eman Ayyub, Russell C Petter
ABSTRACT
Piperazine derivatives of 2-furanyl[1,2,4]triazolo[1,5-a][1,3,5]triazine have recently been shown to be potent and selective adenosine A(2a) receptor antagonists. We now demonstrate that potent and selective A(2a) receptor antagonists could still be obtained when the arylpiperazines are separated from the triazolotriazine core structure by an ethylenediamine spacer. Selected analogs bearing this triazolotriazine or the related triazolopyrimidine core structure have been found to be orally active in a mouse catalepsy model of Parkinson's disease.
Sigma-Aldrich
Aminoacetaldehyde dimethyl acetal, 99%