MilliporeSigma
  • Trisubstituted and tetrasubstituted pyrazolines as a novel class of cell-growth inhibitors in tumor cells with wild type p53.
Trisubstituted and tetrasubstituted pyrazolines as a novel class of cell-growth inhibitors in tumor cells with wild type p53.
Bioorganic & medicinal chemistry (2013-10-22)
Mohammad Abdel-Halim, Adam B Keeton, Evrim Gurpinar, Bernard D Gary, Simon M Vogel, Matthias Engel, Gary A Piazza, Frank M Boeckler, Rolf W Hartmann, Ashraf H Abadi
ABSTRACT
Derivatives with scaffolds of 1,3,5-tri-substituted pyrazoline and 1,3,4,5-tetra-substituted pyrazoline were synthesized and tested for their inhibitory effects versus the p53(+/+) HCT116 and p53(-/-) H1299 human tumor cell lines. Several compounds were active against the two cell lines displaying IC50 values in the low micromolar range with a clearly more pronounced effect on the p53(+/+) HCT116 cells. The compound class shows excellent developability due to the modular synthesis, allowing independent optimization of all three to four key substituents to improve the properties of the molecules.