MilliporeSigma
  • Identification and characterization of small molecules as potent and specific EPAC2 antagonists.
Identification and characterization of small molecules as potent and specific EPAC2 antagonists.
Journal of medicinal chemistry (2013-01-05)
Haijun Chen, Tamara Tsalkova, Oleg G Chepurny, Fang C Mei, George G Holz, Xiaodong Cheng, Jia Zhou
ABSTRACT
EPAC1 and EPAC2, two isoforms of exchange proteins directly activated by cAMP (EPAC), respond to the second messenger cAMP and regulate a wide variety of intracellular processes under physiological and pathophysiological circumstances. Herein, we report the chemical design, synthesis, and pharmacological characterization of three different scaffolds (diarylsulfones, N,N-diarylamines, and arylsulfonamides) as highly potent and selective antagonists of EPAC2. Several selective EPAC2 antagonists have been identified including 20i (HJC0350), which has an apparent IC(50) of 0.3 μM for competing with 8-NBD-cAMP binding of EPAC2 and is about 133-fold more potent than cAMP. Compounds 1 (ESI-05), 14c (HJC0338), and 20i, selected from each series, have exhibited no inhibition of EPAC1-mediated Rap1-GDP exchange activity at 25 μM, indicating that they are EPAC2-specific antagonists. Moreover, live-cell imaging studies using EPAC1, EPAC2, or PKA FRET sensor also demonstrate that 20i functions as an EPAC2 specific antagonist.
Sigma-Aldrich
ESI-05, ≥98% (HPLC)