MilliporeSigma
  • High-dose statin treatment does not alter plasma marker for brain cholesterol metabolism in patients with moderately elevated plasma cholesterol levels.
High-dose statin treatment does not alter plasma marker for brain cholesterol metabolism in patients with moderately elevated plasma cholesterol levels.
Journal of clinical pharmacology (2006-07-01)
Karin M Thelen, Reijo Laaksonen, Hannu Päivä, Terho Lehtimäki, Dieter Lütjohann
ABSTRACT
Statins inhibit endogenous cholesterol synthesis, up-regulate low-density lipoprotein (LDL) receptor expression in mammalian liver cells, and thus decrease circulating LDL-cholesterol concentrations. As cholesterol seems to play a role in the development of neurodegenerative diseases, it is of interest to evaluate the effect of high dosages of statins (eg, atorvastatin or simvastatin) on brain cholesterol metabolism. Plasma samples from 44 participants (aged 30-69 years, 16 men and 18 women) of an earlier randomized, placebo-controlled, double-blind trial, who took 40 mg atorvastatin or 80 mg simvastatin daily for 2 months, were used to analyze total cholesterol, its precursor lathosterol, and its metabolites 24(S)-hydroxycholesterol and 27-hydroxycholesterol. Despite a significant decrease in absolute plasma concentrations of oxysterols, total cholesterol, and its endogenous synthesis rate, indicated by a decreased ratio of lathosterol to cholesterol, the plasma 24(S)-hydroxycholesterol to cholesterol ratio, a surrogate marker of brain cholesterol homeostasis, remained unchanged. Short-term high-dose atorvastatin and simvastatin treatment does not seem to influence brain cholesterol metabolism in patients with moderately elevated plasma cholesterol levels.
Sigma-Aldrich
Cholesteryl palmitate, ≥98% (HPLC; detection at 205 nm)