MilliporeSigma
  • Highly sensitive hepatitis B surface antigen detection by measuring stable nitroxide radical formation with ESR spectroscopy.
Highly sensitive hepatitis B surface antigen detection by measuring stable nitroxide radical formation with ESR spectroscopy.
Free radical biology & medicine (1998-12-05)
T Matsuo, H Shinzawa, H Togashi, M Aoki, K Sugahara, K Saito, T Saito, T Takahashi, I Yamaguchi, M Aoyama, H Kamada
ABSTRACT
In areas where hepatitis B virus (HBV) is prevalent, HBV carriers negative for hepatitis B surface antigen (HbsAg) by enzyme-linked immunosorbent assay (ELISA) have been reported. Moreover, even after screening donor blood for HbsAg and hepatitis B core antibody (HBcAb), post-transfusion hepatitis B continues to occur, though with a decreasing frequency. Therefore, screening tests far more sensitive for detecting HBsAg than those currently available are needed. We developed a highly sensitive method for HBsAg detection. It is based on the recognition of peroxidase activity through measuring the formation of stable nitroxide radical with electron spin resonance (ESR) spectroscopy in the presence of hydrogen peroxide, p-acetamidophenol (p-AP), and 4-hydrazonomethyl-1-hydroxy-2,2,5,5,-tetramethyl-3-imidazoline-3-o xide (HHTIO). A cut-off value was established by testing of 186 healthy adults and 50 HBsAg-positive individuals. The signal to noise (S/N) ratio of less than 1.488 obtained by ESR spectroscopy was considered to be negative and more than 2.181, positive. The p-AP/HHTIO method was found to be 10 times more sensitive than the standard ELISA and reproducibility was excellent. Additional investigations were made on the HBsAg levels in the serum from 26 healthy subjects, in whom cut-off index levels on ELISA were negative but relatively high (range: 0.6 to 1.0); and on 15 patients with non B non C hepatitis. Three of 26 cases and 3 of 15 with non B non C hepatitis were judged to be HBsAg positive. Of these, 5 were found to be positive for HBV DNA by polymerase chain reaction (PCR). It was shown in this study that the p-AP/HHTIO method is practical and useful in screening HBV carriers because of the sensitivity in HBsAg detection, which is comparable to PCR analysis.