MilliporeSigma
  • Demethylation of dimethylsulfoniopropionate to 3-mercaptopropionate by an aerobic marine bacterium.
Demethylation of dimethylsulfoniopropionate to 3-mercaptopropionate by an aerobic marine bacterium.
Applied and environmental microbiology (1994-12-01)
P T Visscher, B F Taylor
ABSTRACT
A bacterium, strain BIS-6, that grew aerobically on dimethylsulfoniopropionate (DMSP) was isolated from an intertidal mud sample. Strain BIS-6 quantitatively demethylated DMSP and 3-methiolpropionate to 3-mercaptopropionate. Strain BIS-6 was a versatile methylotroph growing on the osmolytes DMSP and glycine betaine and their methylated degradation products (dimethyl glycine, sarcosine, methylamines, and dimethyl sulfide.
Sigma-Aldrich
Methyl 3-(methylthio)propionate, 98%