MilliporeSigma
  • Cyclosporine triggers endoplasmic reticulum stress in endothelial cells: a role for endothelial phenotypic changes and death.
Cyclosporine triggers endoplasmic reticulum stress in endothelial cells: a role for endothelial phenotypic changes and death.
American journal of physiology. Renal physiology (2008-11-07)
Nicolas Bouvier, Jean Pierre Flinois, Jerome Gilleron, François-Ludovic Sauvage, Christophe Legendre, Philippe Beaune, Eric Thervet, Dany Anglicheau, Nicolas Pallet
ABSTRACT
Calcineurin inhibitors cyclosporine and tacrolimus are effective immunosuppressants, but both substances have the same intrinsic nephrotoxic potential that adversely affects allograft survival in renal transplant patients and causes end-stage renal disease in other solid organ or bone marrow transplant recipients. Endothelial cells are the first biological interface between drugs and the kidney, and calcineurin inhibitors may influence endothelial function and viability in a number of ways. Notably, endothelial cells have recently been shown to contribute to the accumulation of interstitial fibroblasts in nonrenal models, through endothelial-to-mesenchymal transition. Here we demonstrate that cyclosporine, but not tacrolimus or its metabolites, induces morphological and phenotypic endothelial changes suggestive of a partial endothelial-to-mesenchymal transition in human umbilical arterial endothelial cells. We identify for the first time a contingent of interstitial myofibroblasts that coexpress endothelial markers in rat kidneys treated with cyclosporine, suggesting that endothelial-to-mesenchymal transition could occur in vivo. Finally, our findings suggest that endoplasmic reticulum stress triggered by cyclosporine induces endothelial cells to undergo endothelial phenotypic changes suggestive of a partial endothelial-to-mesenchymal transition, whereas salubrinal partially preserves the endothelial phenotype. Inversely, tacrolimus does not induce endothelial-to-mesenchymal transition or endoplasmic reticulum stress. In conclusion, this study demonstrates for the first time that cyclosporine, and not tacrolimus, induces endoplasmic reticulum stress in endothelial cells. Our findings also suggest that endoplasmic reticulum stress contributes to endothelial cell death and phenotypic changes similar to a partial endothelial-to-mesenchymal transition.