MilliporeSigma
  • Modulation of the cell kinetics of pig skin by the topical application of evening primrose oil or Lioxasol.
Modulation of the cell kinetics of pig skin by the topical application of evening primrose oil or Lioxasol.
Cell proliferation (1997-08-01)
G M Morris, J W Hopewell, M Harold, G A Ross, N M Nadejina, I Gusev, I Flockhart
ABSTRACT
The daily topical application of two compounds, a cream containing 10% evening primrose oil (EPO) and Lioxasol (a compound used clinically to treat radiation burns), resulted in increased cell proliferative activity in the skin of female Large White pigs. The effect was most pronounced in the case of the EPO based cream, and was comparable in magnitude with that observed in a previous study on pig skin using orally administered EPO. There was an increase in the size of the rete pegs in the epidermis by 6 weeks after the start of application of the EPO cream. However, this did not translate into an increase in the total thickness of the viable epidermis (excluding the stratum corneum) due to a reduction in the density of rete pegs, from 2 weeks after treatment. Lioxasol had no overall effect on the size of the rete pegs. The labelling index (LI) of cells in the basal layer of the epidermis of pigs receiving a daily topical application of EPO increased progressively with time from the start of application. The LI was maximal (17.9 +/- 2.4%) at the end of the observation period (8 weeks) at which time it was a factor of approximately 2 higher than in the basal layer prior to treatment. A considerably less marked increase in the LI of the basal layer was seen after the application of Lioxasol. The overall increase was approximately 20%, relative to the LI in the untreated epidermis. Labelled cell nuclei were also counted in the papillary dermis. After the application of the EPO cream, no significant increase in the number of labelled cells was observed until week 8, at which time values were approximately twice those in untreated skin. In Lioxasol treated skin the effect on the numbers of labelled cells in the papillary dermis was more immediate, with a approximately 60% increase at 2 weeks. This enhanced level of labelling was maintained until the end of the observation period of 10 weeks. Studies on the cell kinetics of the skin using the alcohol component of the Lioxasol preparation suggested that alcohol rather than Lioxasol was the most significant ingredient. It was concluded that the EPO cream merited further evaluation as a potential modulator of skin response to ionizing radiation.
Sigma-Aldrich
2-Allyloxyethanol, 98%
Pricing and availability is not currently available.