MilliporeSigma
[Interaction of L-glutamic acid with human T-lymphocytes].
Bioorganicheskaia khimiia (1998-03-07)
I A Kostanyan, E V Navolotskaya, R I Nurieva, V P Zav'ialov, V M Lipkin
ABSTRACT
A specific interaction of [3H]Glu with T lymphocytes from the blood of healthy donors (Kd = 0.236 microM) was revealed and described. It was found that unlabeled quisqualate, a structural analogue of L-glutamic acid, and unlabeled dipeptides Ala-Glu, Glu-Ala, and Glu-Glu competitively inhibit the specific binding of [3H]Glu to T lymphocytes (with Ki 0.19, 2.4, 3.4, and 1.2 microM, respectively). Binding experiments with conjugates of labeled and unlabeled glutamic acid with dextran showed that the receptors of [3H]Glu are localized on the outer surface of the plasma membrane of T lymphocytes.
Sigma-Aldrich
Glu-Glu