MilliporeSigma
  • Synthesis, structure, dopamine transporter affinity, and dopamine uptake inhibition of 6-alkyl-3-benzyl-2-[(methoxycarbonyl)methyl]tropane derivatives.
Synthesis, structure, dopamine transporter affinity, and dopamine uptake inhibition of 6-alkyl-3-benzyl-2-[(methoxycarbonyl)methyl]tropane derivatives.
Journal of medicinal chemistry (1998-01-22)
S A Lomenzo, S Izenwasser, J L Katz, P D Terry, N Zhu, C L Klein, M L Trudell
ABSTRACT
A series of 6-alkyl-3 beta-benzyl-2-[(methoxycarbonyl)methyl]tropane analogues were synthesized and evaluated as cocaine binding site ligands at the dopamine transporter (DAT). The in vitro affinity (Ki) for the DAT of the 6-alkyl-3 beta-benzyl-2-[(methoxycarbonyl) methyl]tropane analogues was determined by inhibition of [3H]WIN 35,428 in rat caudate putamen tissue. The inhibition of dopamine uptake (IC50) was also measured for selected compounds which demonstrated moderate affinity for the dopamine transporter. The unsubstituted enantiopure analogues (-)-19a (Ki = 33 nM) and surprisingly (+)-20a (Ki = 60 nM) were found to be almost equipotent with the high-affinity binding components of cocaine and WIN 35,065-2 and exhibited slightly more potent dopamine uptake inhibition than both cocaine and WIN 35,065-2. In general, substitution at the 6-position of racemic 19a and 20a with alkyl groups was found to result in decreased activity relative to increased chain length of the substituent. The 3 beta-benzyl-2 beta-[(methoxycarbonyl)methyl]-6 beta-methyltropane (21b; Ki = 57 nM) was the only 6-alkyl derivative to exhibit moderately potent activity. The 6 beta-isomer 21b was 4-fold more potent than the 6 alpha-isomer 19b (Ki = 211 nM) and was nearly equipotent with (-)-19a and (+)-20a as well as with cocaine and WIN 35,065-2. The results of this study further demonstrate the steric constraints associated with the C(6)-C(7) methylene bridge of the tropane ring system for molecular recognition of cocaine analogues at the cocaine binding site(s) on the DAT.