MilliporeSigma
  • Enhancement or induction of neurite formation by a protein tyrosine phosphatase inhibitor, 3,4-dephostatin, in growth factor-treated PC12h cells.
Enhancement or induction of neurite formation by a protein tyrosine phosphatase inhibitor, 3,4-dephostatin, in growth factor-treated PC12h cells.
Biochemical and biophysical research communications (1997-09-23)
S Fujiwara, T Watanabe, T Nagatsu, J Gohda, M Imoto, K Umezawa
ABSTRACT
We studied the effect of the 3,4-dihydroxy analogue of dephostatin (3,4-dephostatin), an inhibitor of protein-tyrosine phosphatase (PTPase), on the differentiation of rat pheochromocytoma PC12 cells. 3,4-Dephostatin accelerated NGF-induced neurite formation in PC12h cells, a subline of PC12 cells, whereas the inhibitor alone did not induce neurite formation. It sustained the NGF-induced tyrosine phosphorylation of several proteins, most prominently that of mitogen-activated protein (MAP) kinase. EGF alone did not induce differentiation in PC12h cells, but it induced neurite formation in the presence of 3,4-dephostatin. The inhibitor also prolonged EGF-induced tyrosine phosphorylation and activation of MAP kinase. An inactive analogue of dephostatin, 2'-O-methyl-dephostatin showed no effect on either neurite formation or MAP kinase tyrosine phosphorylation in NGF or EGF-treated PC12h cells. Thus, we demonstrated that the PTPase inhibitor could enhance growth factor-induced differentiation in PC12 cells possibly by sustaining the MAP kinase activity.
Sigma-Aldrich
Methyl-3,4-dephostatin, >98% (HPLC), powder