MilliporeSigma
Opioid analgesics.
British journal of hospital medicine (1997-02-05)
A R Rushton, J R Sneyd
ABSTRACT
This article discusses recent developments in opioid pharmacology. Selective activity at different opioid receptors promises, but has not yet provided, a strong analgesic without opioid side-effects. The startling pharmacokinetic profile of the new esterase-metabolized intravenous opioid, remifentanil, is reflected in the rapid onset of effect and short, predictable recovery, independent of duration of infusion.
Sigma-Aldrich
Meptazinol hydrochloride