MilliporeSigma
  • Determination of haloperidol and its reduced metabolite in human plasma by liquid chromatography-mass spectrometry with electrospray ionization.
Determination of haloperidol and its reduced metabolite in human plasma by liquid chromatography-mass spectrometry with electrospray ionization.
Journal of chromatography. B, Biomedical sciences and applications (1997-01-24)
H Hoja, P Marquet, B Verneuil, H Lotfi, J L Dupuy, B Pénicaut, G Lachâtre
ABSTRACT
A sensitive and accurate liquid chromatographic-electrospray mass spectrometric (LC-ES-MS) method for the determination of haloperidol (H) and reduced haloperidol (RH) in human plasma is presented, using chlorohaloperidol as the internal standard. A 2-ml volume of plasma was subjected to basic (NaOH) extraction, acid (HCl) back-extraction, acid wash and basic (NaOH) re-extraction. The extraction solvent was hexane-isoamyl alcohol (99:1, v/v) for the whole procedure. A Nucleosil C18 column (150 x 1 mm) was used for high-performance liquid chromatography, together with 2 mM HCOONH4-acetonitrile (55:45, v/v; pH 3.0) as the mobile phase. For each drug, four characteristic ions were monitored. Linearity was assessed in the ranges 0.1-50 and 0.25-50 ng/ml for H and RH, respectively. Recoveries were 58 and 70% and detection limits were 0.075 and 0.100 ng/ml for H and RH, respectively. Correlation coefficients were better than 0.999 for both compounds. R.S.D.s for repeatability and reproducibility at 0.25 ng/ml were 11.1 and 8.5% for H and 9.4 and 11.2% for RH, respectively. One of the main advantages of (LC-ES-MS) over other detection systems is the increase in selectivity obtained by monitoring three ions of confirmation for each of the drugs.
Product Description
Supelco
Haloperidol metabolite II, analytical standard
Pricing and availability is not currently available.