MilliporeSigma
  • Suppression of the voltage-gated K+ current of human megakaryocytes by thrombin and prostacyclin.
Suppression of the voltage-gated K+ current of human megakaryocytes by thrombin and prostacyclin.
Biochimica et biophysica acta (1997-03-01)
L Kapural, A Fein
ABSTRACT
We examined the effects of platelet activators and inhibitors of platelet function on the voltage-gated delayed rectifier K+ current of human megakaryocytes. We found that both the activators such as thrombin, the thrombin receptor peptide (TRP42-47) and ADP and the inhibitors such as prostacyclin suppressed the delayed rectifier current through two different mechanisms. The cAMP dependent protein kinase (A-kinase) inhibitor IP20 blocked the suppression of the delayed rectifier current by prostacyclin and failed to block the suppression by thrombin, TRP42-47 and ADP. The effects of IP20 suggest that the action of prostacyclin is mediated by A-kinase and the action of the three activators is not mediated by A-kinase. Pertussis toxin (PTX) an inhibitor of the inhibitory GTP-binding proteins (Gi) blocked the suppression of the delayed rectifier current by thrombin, TRP42-47 and ADP and failed to block the suppression by prostacyclin. The effects of PTX suggests that the action of the three activators is mediated by Gi or some other PTX-sensitive GTP-binding protein. We speculate that thrombin and other platelet activators that activate Gi may be suppressing the delayed rectifier current via a direct interaction of Gi or a subunit of it with the delayed rectifier potassium channel itself.