MilliporeSigma
  • Effect of three galanin antagonists on the pressor response to galanin in the Cane toad, Bufo marinus.
Effect of three galanin antagonists on the pressor response to galanin in the Cane toad, Bufo marinus.
Regulatory peptides (1996-12-17)
D A Mahns, G P Courtice
ABSTRACT
Galanin is a neuropeptide that causes a marked pressor response in several non-mammalian vertebrate species, and some marsupials. In this study, the effect of three galanin antagonists were tested on the pressor response to an intravenous dose (6.3 nmol/kg) of porcine galanin in anaesthetised Cane toads, Bufo marinus. Antagonists were injected at either 20 or 50 times the molar dose (x MD) of galanin. The antagonist, C7 (Galanin 1-13-spantide) reduced the pressor effect of galanin by 32.2 +/- 6.0% when delivered at 20 x MD (n = 4) and by 42.9 +/- 15.7% when delivered at 50 x MD (n = 4) of galanin, the response recovering within 30 min. A second antagonist, M32a (Galanin 1-13-NPY 24-36) had no effect on the pressor response to galanin at 20 x MD (n = 4), but significantly reduced the pressor effect by 54.8 +/- 6.4% at 50 x MD (n = 5), which also recovered within 30 min. Administration of a third antagonist, galantide or M15 (Galanin 1-13-Substance P5-11), resulted in a profound drop in blood pressure, and did not affect the response to galanin at either dose. In conclusion, C7 and M32a are effective, short-term antagonists of the blood pressure effects of galanin in the toad.