MilliporeSigma
  • High-performance liquid chromatographic method for the determination of the anthelmintic nitroxynil in cattle muscle tissue with on-line anion-exchange clean-up.
High-performance liquid chromatographic method for the determination of the anthelmintic nitroxynil in cattle muscle tissue with on-line anion-exchange clean-up.
Journal of chromatography (1993-04-02)
J A Tarbin, G Shearer
ABSTRACT
A high-performance liquid chromatographic method for the determination of the anthelmintic nitroxynil has been developed. The drug was extracted from cattle muscle tissue with 1% triethylamine in acetonitrile. The extract was evaporated to dryness and taken up in 0.1 M ammonium acetate-acetonitrile (50:50, v/v). The extract was then injected onto a polymeric anion-exchange precolumn. After clean-up with 0.1 M ammonium acetate-acetonitrile (50:50, v/v) for 5 min, the precolumn was eluted with 1% aqueous trifluoroacetic acid-acetonitrile (50:50, v/v) onto a PLRP-S polymer column and chromatographed with a mobile phase of 0.01 M phosphate pH 7-acetonitrile (80:20, v/v). Detection was by ultraviolet at 273 nm. Average recoveries at four levels from 0.005 to 1.000 mg kg-1 were > 88%. The limit of determination was 0.005 mg kg-1.
Product Description
Supelco
Nitroxinil, VETRANAL®, analytical standard