MilliporeSigma
  • The unique steroidogenesis of the aldosteronoma in the differential diagnosis of primary aldosteronism.
The unique steroidogenesis of the aldosteronoma in the differential diagnosis of primary aldosteronism.
The Journal of clinical endocrinology and metabolism (1993-04-01)
S Ulick, J D Blumenfeld, S A Atlas, J Z Wang, E D Vaughan
ABSTRACT
18-Hydroxycortisol and 18-oxocortisol have been isolated from the urine of patients with aldosterone producing adrenocortical adenomas, but not from those with idiopathic hyperaldosteronism associated with bilateral adrenal hyperplasia. These C-18 oxygenated cortisols are biosynthesized by the substitution of cortisol for the normal substrate, corticosterone, in the terminal oxidase system required for the biosynthesis of 18-hydroxycorticosterone and aldosterone. To make use of this biochemical difference between the two groups in the preoperative diagnosis of primary aldosteronism, we have developed and utilized a specific primary standard analytical method, stable isotope dilution mass fragmentography, for quantifying 18-hydroxycortisol and the tetrahydro metabolite of 18-oxocortisol in 24-h urine samples. The normal range by this technique of 4.6 +/- 1.8 micrograms/day tetrahydro 18-oxocortisol and 43 +/- 23 micrograms/day 18-hydroxycortisol in urine was lower and narrower than previous estimates using other methods. Excretion of the 18-oxocortisol metabolite ranged from 2-12 micrograms/day in bilateral hyperplasia and 17-1203 micrograms/day in typical adenomas. 18-Hydroxycortisol excretion similarly separated bilateral hyperplasia (23-59 micrograms/day) from typical adenomas (60-2750 micrograms/day). The cortisol C-18 oxidation pathway describes a unique steroidogenic mechanism in the aldosteronoma not present in idiopathic aldosteronism due to bilateral adrenal hyperplasia and as such provides a basis for the biochemical classification of primary aldosteronism and the differentiation of these two groups. This unique biochemistry was also observed in unilateral hyperplasia but not in the renin-dependent aldosteronoma.
Sigma-Aldrich
3α,5β-Tetrahydroaldosterone, ≥98% (CP)