MilliporeSigma
  • Creatinine assay by the fuller's earth procedure or by enzymatic determination is adequate for urine but not plasma of mice.
Creatinine assay by the fuller's earth procedure or by enzymatic determination is adequate for urine but not plasma of mice.
Comparative biochemistry and physiology. B, Comparative biochemistry (1993-11-01)
M H Meyer, S Smith, R A Meyer
ABSTRACT
1. Mouse plasma creatinine measured by fuller's earth (15.1 +/- 0.5(41) microM, mean +/- SE) exceeded the same pools read by HPLC [7.5 +/- 0.2(41); P < 0.001]. 2. For mouse urine, the fuller's earth method (3.26 +/- 0.21 mM) slightly exceeded the HPLC method (2.65 +/- 0.18; P < 0.001) with good correlation (r = 0.915, P < 0.001). 3. An enzymatic method (creatinine PAP, Boehringer-Mannheim Diagnostics) for plasma creatinine (5.2 +/- 0.3 microM) was equivalent to HPLC (6.1 +/- 0.1, NS) but with poor correlation (r = 0.23, NS). 4. Creatinine in the urine agreed closely between HPLC (2.84 +/- 0.12 mM) and the enzymatic procedure (2.95 +/- 0.17, NS) with good correlation (r = 0.845, P < 0.001).
Sigma-Aldrich
Fuller′s earth, -100 mesh particle size