MilliporeSigma
  • 17 beta-estradiol glucuronide: an inducer of cholestasis and a physiological substrate for the multidrug resistance transporter.
17 beta-estradiol glucuronide: an inducer of cholestasis and a physiological substrate for the multidrug resistance transporter.
Cancer research (1993-11-15)
M Gosland, C Tsuboi, T Hoffman, S Goodin, M Vore
ABSTRACT
The multidrug resistance (MDR) gene family has been shown to be highly expressed in several normal tissues including the canalicular membrane of the hepatocyte. We report that a cholestatic estrogen metabolite, 17 beta-estradiol glucuronide (E217G), is a substrate for the MDR transporter, P-glycoprotein. In cytotoxicity studies, the MDR sarcoma cell line Dx5 was 4.7-fold resistant to E217G, and the K562/R7 leukemia MDR cell line was 5.0-fold resistant to E217G relative to their parental cell lines. There was also a 2- to 3-fold accumulation defect of [3H]E217G in the MDR cells relative to their parental cell lines. E217G (100 microM) modulated resistance ot doxorubicin, taxol, vinblastine, and etoposide in the Dx5 cells, completely reversing the 30- to 60-fold resistance observed with these agents. E217G had no effect on the toxicity of these compounds in the parental cell line (MES-SA). In contrast, MDR cells were not resistant to the noncholestatic estrogen metabolite, estriol 3-glucuronide, and this metabolite did not modulate resistance to MDR substrates. ATP-dependent transport of [3H]E217G in rat canalicular membranes was inhibited by several MDR substrates including vinblastine, etoposide, verapamil, cyclosporine, and PSC-833.