MilliporeSigma
  • Inhibitory effects of benzyl isothiocyanate and benzyl thiocyanate on diethylnitrosamine-induced hepatocarcinogenesis in rats.
Inhibitory effects of benzyl isothiocyanate and benzyl thiocyanate on diethylnitrosamine-induced hepatocarcinogenesis in rats.
Japanese journal of cancer research : Gann (1993-08-01)
S Sugie, A Okumura, T Tanaka, H Mori
ABSTRACT
The effects of two aromatic thiocyanates, benzyl isothiocyanate (BITC) and benzyl thiocyanate (BTC), on diethylnitrosamine (DEN)-induced hepatocarcinogenesis were examined in rats. A total of 108 male ACI/N rats, 5 weeks old, were divided into 6 groups (18 rats in each). Group 1 was given a single i.p. injection of DEN (200 mg/kg body weight) one week after the start of the experiment and then kept on the basal diet until the end of the experiment (1 year). Groups 2 and 3 were treated with DEN and received dietary BITC (100 ppm) or BTC (100 ppm), respectively, throughout the experimental duration. Groups 4 and 5 were not given the carcinogen and were fed the diet containing BITC or BTC, respectively. Group 6 was kept on the basal diet alone and served as a control. Liver neoplasms were seen in Groups 1, 2 and 3. Incidence and average number of liver neoplasms in Group 2 were significantly smaller than in Group 1 (P < 0.0005 and P < 0.001, respectively). The incidence of liver neoplasms in Group 3 was slightly lower than in Group 1, although the difference was not statistically significant. The numbers of glutathione S-transferase placental form (GST-P)-positive foci in Group 2 and gamma-glutamyltranspeptidase (GGT)-positive foci in Groups 2 and 3 were significantly smaller than those in Group 1 (P < 0.001). The average and unit areas of GST-P- or GGT-positive foci in Group 2 or 3 were also significantly smaller than those in Group 1 (P < 0.05). These results suggest that BITC and BTC are chemopreventive agents for DEN-induced liver tumorigenesis.
Sigma-Aldrich
Benzyl thiocyanate, ≥95.0% (GC)