MilliporeSigma
  • Neurochemical characterization of the alterations in the noradrenergic afferents to the cerebellum of adult rats exposed to X-irradiation at birth.
Neurochemical characterization of the alterations in the noradrenergic afferents to the cerebellum of adult rats exposed to X-irradiation at birth.
Journal of neurochemistry (1993-08-01)
A M Dopico, L M Zieher
ABSTRACT
A single dose of x-irradiation was applied on the cephalic end of newborn rats, and the alterations in the noradrenergic afferents to the cerebellum were studied 180 days later. A net increase in the noradrenaline content of cerebellum was found (122% of nonirradiated controls). The response of noradrenaline content to reserpine injection (0.9 mg/kg, i.p.) was similar in exposed and control rats. Likewise, the 3H release induced by Ro 4-1284 from cerebellar cortex slices labeled with [3H]noradrenaline was unmodified by x-rays, although a mild increase in the spontaneous efflux of 3H was found. The retention of 3H by the slices was reduced in exposed animals (58% of controls). Both the in vitro activity of tyrosine hydroxylase and the accumulation of L-3,4-dihydroxyphenylalanine (L-DOPA) were not significantly different between x-treated rats and controls. In contrast, monoamine oxidase activity was markedly reduced in x-irradiated cerebellum (38% of controls). The x-ray-induced decrease in cerebellar weight (-60%) resulted in marked increases in noradrenaline concentration (223%), tyrosine hydroxylase activity per milligram of protein (206%), and 3H retention (50%). The accumulation of L-DOPA per gram of tissue was also increased at every time considered. These data indicate that x-irradiation at birth produces a cerebellar loss not completely shared by the noradrenergic afferents, and a permanent imbalance between the noradrenergic afferent input and its target cells might eventually result.(ABSTRACT TRUNCATED AT 250 WORDS)