MilliporeSigma
  • The effects of GR79236 on plasma fatty acid concentrations, heart rate and blood pressure in the conscious rat.
The effects of GR79236 on plasma fatty acid concentrations, heart rate and blood pressure in the conscious rat.
European journal of pharmacology (1994-05-12)
C J Gardner, D J Twissell, J Coates, P Strong
ABSTRACT
GR79236 (N-[(1S,trans)-2-hydroxycyclopentyl]adenosine) is an orally active adenosine A1 receptor agonist, which decreases plasma non-esterified fatty acid levels in fasted rats. This study has quantified the effects of GR79236 on plasma non-esterified fatty acid levels, blood pressure and heart rate in conscious rats. Blood pressure and heart rate were continuously recorded in rats for 30 min before and 1 h after oral dosing with GR79236 (0.03-3 mg/kg) or vehicle. Plasma non-esterified fatty acid concentrations were determined in blood sampled before and 1 h after dosing. GR79236 produced dose-related reductions in plasma non-esterified fatty acid concentrations, with an almost maximal effect (63.2%) occurring after 1 mg/kg. This dose induced a small but significant decrease in heart rate (12.0%), and a non-significant decrease in mean blood pressure (6.3%). Thus, in the conscious rat, GR79236 can exert profound antilipolytic effects with minimal effects on blood pressure and heart rate.