MilliporeSigma
  • Digitalis-like compounds in the toad Bufo viridis: interactions with plasma proteins.
Digitalis-like compounds in the toad Bufo viridis: interactions with plasma proteins.
Journal of cardiovascular pharmacology (1993-01-01)
D Lichtstein, I Gati, H Ovadia
ABSTRACT
Digitalis-like compounds (DLC), normal constituents of animal tissues, are possible regulators of the Na+,K(+)-ATPase implicated in water and salt homeostasis. DLC are present in toad (Bufo viridis) tissues. Although DLC highest levels were found in toad skin, it was also detected in plasma and many internal organs. The abundant distribution and the different levels of DLC in various tissues exclude the possibility that toxicity is the only function of these compounds in the toad. The concentration of DLC in toad plasma is 30 microM, out of which 25-30% is bound to plasma proteins. Fractionation of toad plasma proteins on a G-100 Sephadex column followed by the extraction of DLC from the plasma proteins revealed that DLC are bound primarily to proteins of 48,000-53,000 Da. These results establish the existence of bufodienolide-binding protein(s) in animal plasma.