- Effect of anilazine on growth, respiration and enzyme activity of Escherichia coli.
Effect of anilazine on growth, respiration and enzyme activity of Escherichia coli.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes (1981-01-01)
F C Garg, P Tauro, M M Mishra, R K Grover
PMID7034397
ABSTRACT
The effect of anilazine on growth, respiration and enzyme activity of E. coli has been studied. Anilazine delays the growth of E. coli by prolonging the lag period. It inhibits glucose oxidation by 60% and succinate oxidation by 100%. It also inhibits in vitro succinic dehydrogenase activity. It seems that the inhibition of E. coli by anilazine is because of inhibition of respiratory enzymes.
MATERIALS