MilliporeSigma
  • Placental transfer and renal clearance of sulphamethoxazole and its metabolite N4-acetylsulphamethoxazole in a pregnant ewe.
Placental transfer and renal clearance of sulphamethoxazole and its metabolite N4-acetylsulphamethoxazole in a pregnant ewe.
Journal of veterinary pharmacology and therapeutics (1983-03-01)
T B Vree, J J Reekers-Ketting, J F Nouws, T H Arts
ABSTRACT
Sulphamethoxazole and its metabolite, N4-acetylsulphamethoxazole, were shown to cross the placenta of a pregnant ewe. The plasma concentration of N4-acetylsulphamethoxazole increased in the foetus due to the limited elimination properties of the immature kidney. The amniotic fluid concentration of sulphamethoxazole and N4-acetylsulphamethoxazole were low, amounting to 2.0 and 0.46 micrograms/ml respectively. Protein binding of the sulphonamides was slightly higher in the foetus than in the ewe.
Product Description
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard