MilliporeSigma
Conformational influences of N2-methylation in chemotactic peptides.
International journal of peptide and protein research (1984-11-01)
P W Jeffs, S L Heald, D F Chodosh, D S Eggleston
ABSTRACT
Physical and structural data on two of a series of related N-formyl methionine peptides are reported. The effects of peptide bond N-methylation on chemotactic response and on conformational properties observed in solution, in the solid state and by conformational energy calculations are described.