MilliporeSigma
Membership mutation of the genetic code: loss of fitness by tryptophan.
Proceedings of the National Academy of Sciences of the United States of America (1983-10-01)
J T Wong
ABSTRACT
Bacillus subtilis strain QB928, a tryptophan-auxotroph, was serially mutated to yield strain HR15. For QB928, tryptophan functioned as a competent amino acid and 4-fluorotryptophan as merely an inferior analogue. For HR15, these roles were reversed. The tryptophan/4-fluorotryptophan growth ratio decreased by a factor of 2 X 10(4) in the transition from QB928 to HR15.