MilliporeSigma
A controlled trial of cinromide.
Epilepsia (1983-08-01)
J W Allen, M J Hamilton, J Oxley, A Richens
ABSTRACT
A double-blind controlled trial of cinromide in 25 adult subjects with refractory epilepsy using the maximum tolerated dose showed that it possessed no significant antiepileptic properties.